Learn More
Bioworld GST-Tagged Protein Purification Buffer Kit, 200 mL
Supplier:  Bioworld 208400432
Encompass_Preferred
bio-World's GST-Tagged Protein Purification Buffer Kit consists of a set of buffers necessary for OnePass™ GST-Tagged Recombinant Protein Purification System(Glutathione-Separopore conveniently prepacked spin columns for gravity flow chromatography) or batch purification using Glutathione Separopore.Kit Contents:• GST Binding Buffer (100 ml)• Wash Buffer (200ml)• GST Elution Buffer (50ml)
Catalog No. 50-198-486