Learn More
Proteintech Group Inc GST-Trap Agarose, kit
SDP
Supplier:  Proteintech Group Inc STAK20
Encompass
GST-Trap is an affinity resin for IP of GST fusion proteins. It consists of a GST Nanobody/ VHH coupled to agarose beads.
Catalog No. 50-225-8868