Learn More
Aviva Systems Biology Corp GSTA3 Antibody - middle region (ARP48484_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48484_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GSTA3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GSTA3. Alternate names: GSTA3-3, GTA3, MGC22232ApplicationWB. Species reactivity: Human, Mouse, Sheep
Catalog No. 50-014-53932