Learn More
Aviva Systems Biology Corp GSTA4 Antibody - N-terminal region (ARP48554_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48554_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GSTA4. It was validated on Western Blot. . Alternate names: DKFZp686D21185, GSTA4-4, GTA4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit
Catalog No. 50-285-367