Learn More
Aviva Systems Biology Corp GSTA4 Antibody - C-terminal region (ARP48555_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48555_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GSTA4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GSTA4. Alternate names: DKFZp686D21185, GSTA4-4, GTA4ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59857