Learn More
Aviva Systems Biology Corp GSTA5 Antibody - middle region (ARP49087_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49087_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GSTA5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GSTA5. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-60017