Learn More
Aviva Systems Biology Corp GSTM1 Antibody - N-terminal region (ARP41769_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41769_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GSTM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GSTM1. Alternate names: GST1, GSTM1-1, GSTM1a-1a, GSTM1b-1b, GTH4, GTM1, H-B, MGC26563, MU, MU-1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-57989