Learn More
Aviva Systems Biology Corp GSTM2 Antibody - N-terminal region (ARP41818_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41818_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GSTM2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human GSTM2. Alternate names: GST4, GSTM, GSTM2-2, GTHMUS, MGC117303ApplicationIHC, WB. Species reactivity: Human, Zebrafish
Catalog No. 50-014-58006