Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GSTM2 Human N-His | >95.0% | 500 UG
SDP

Catalog No. 5000292795
Encompass_Preferred

Recombinant full-length human glutathione S-transferase mu 2 (GSTM2) with an N-terminal 6xHis tag, expressed in Escherichia coli and supplied as a lyophilized powder for biochemical and structural studies. The protein corresponds to isoform-1 (M1-K218), has an approximate molecular weight of ~26 kDa, and purity greater than 95% by reducing SDS-PAGE.

  • Full-length isoform-1 (M1-K218)
  • N-terminal 6xHis purification tag
  • Molecular weight approximately 26 kDa
  • Purity greater than 95% as determined by reducing SDS-PAGE
  • Lyophilized powder formulated in Tris-NaCl with trehalose and Tween-80
  • Expressed in Escherichia coli
  • Do not reconstitute below 100 μg/mL in ddH2O
  • Store at -20°C; freeze aliquots at -20°C or -80°C for long-term storage
  • Suitable for enzymatic assays, structural studies, and antibody production

Catalog No. 50-002-92795 Supplier Medchemexpress LLC Supplier No. HYP75152A500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant full-length human glutathione S-transferase mu 2 (GSTM2) with an N-terminal 6xHis tag, expressed in Escherichia coli and supplied as a lyophilized powder for biochemical and structural studies. The protein corresponds to isoform-1 (M1-K218), has an approximate molecular weight of ~26 kDa, and purity greater than 95% by reducing SDS-PAGE.

  • Full-length isoform-1 (M1-K218)
  • N-terminal 6xHis purification tag
  • Molecular weight approximately 26 kDa
  • Purity greater than 95% as determined by reducing SDS-PAGE
  • Lyophilized powder formulated in Tris-NaCl with trehalose and Tween-80
  • Expressed in Escherichia coli
  • Do not reconstitute below 100 μg/mL in ddH2O
  • Store at -20°C; freeze aliquots at -20°C or -80°C for long-term storage
  • Suitable for enzymatic assays, structural studies, and antibody production

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.