Learn More
Medchemexpress LLC GSTM2 protein, human (His) | 1 MG
SDP
Catalog No. 5000292989
Encompass_Preferred
Recombinant human Glutathione S-transferase Mu 2 (GSTM2) expressed in Escherichia coli and supplied with an N-terminal 6xHis tag. Supplied as a purified protein (approx. 26 kDa) for biochemical assays, enzyme activity measurements, and protein-protein interaction studies. Pack sizes include 1 mg.
Catalog No. 50-002-92989 Supplier Medchemexpress LLC Supplier No. HYP75152A1MG
Recombinant human Glutathione S-transferase Mu 2 (GSTM2) expressed in Escherichia coli and supplied with an N-terminal 6xHis tag. Supplied as a purified protein (approx. 26 kDa) for biochemical assays, enzyme activity measurements, and protein-protein interaction studies. Pack sizes include 1 mg.