Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GSTM2 protein, human (His) | >95.0% | 100 UG
SDP

Catalog No. 5000292991
Encompass_Preferred

Recombinant full-length human GSTM2 protein with an N-terminal 6xHis tag, expressed in E. coli and supplied as a lyophilized powder for research use. The protein is suitable for enzymatic assays and protein characterization, offering high purity and measured activity in a stabilized formulation.

  • Full-length human sequence expressed in E. coli.
  • N-terminal 6xHis tag for purification and detection.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Approximate molecular weight 26 kDa.
  • Endotoxin less than 1 EU/μg.
  • Bioactivity 10-15 U/mg using CDNB conjugation assay.
  • Lyophilized formulation with stabilizers; reconstitute at ≥100 μg/mL in ddH2O.
  • Store at -20°C; after reconstitution stable 4°C for 1 week or -20°C longer with carrier protein.

Catalog No. 50-002-92991 Supplier Medchemexpress LLC Supplier No. HYP75152A100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant full-length human GSTM2 protein with an N-terminal 6xHis tag, expressed in E. coli and supplied as a lyophilized powder for research use. The protein is suitable for enzymatic assays and protein characterization, offering high purity and measured activity in a stabilized formulation.

  • Full-length human sequence expressed in E. coli.
  • N-terminal 6xHis tag for purification and detection.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Approximate molecular weight 26 kDa.
  • Endotoxin less than 1 EU/μg.
  • Bioactivity 10-15 U/mg using CDNB conjugation assay.
  • Lyophilized formulation with stabilizers; reconstitute at ≥100 μg/mL in ddH2O.
  • Store at -20°C; after reconstitution stable 4°C for 1 week or -20°C longer with carrier protein.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.