Learn More
Medchemexpress LLC Gstm2 protein, human (his, solution) | >95.0% | 5 UG
SDP
Catalog No. 5000291418
Encompass_Preferred
Recombinant human glutathione S-transferase Mu 2 (GSTM2) with an N-terminal 6xHis tag, expressed in Escherichia coli and supplied lyophilized from a Tris-based buffer. The protein is approximately 26 kDa and >95% pure by reducing SDS-PAGE, and is intended for use in biochemical assays, structural studies, and antibody validation.
  • N-terminal 6xHis tag for affinity purification and detection.
  • Expressed in Escherichia coli for high yield production.
  • Lyophilized formulation for extended stability and storage.
  • Purity greater than 95% as assessed by reducing SDS-PAGE.
  • Approximate molecular weight of 26 kDa.
  • Suitable for biochemical assays and antibody validation.
Catalog No. 50-002-91418 Supplier Medchemexpress LLC Supplier No. HYP75152A5UG
Recombinant human glutathione S-transferase Mu 2 (GSTM2) with an N-terminal 6xHis tag, expressed in Escherichia coli and supplied lyophilized from a Tris-based buffer. The protein is approximately 26 kDa and >95% pure by reducing SDS-PAGE, and is intended for use in biochemical assays, structural studies, and antibody validation.
  • N-terminal 6xHis tag for affinity purification and detection.
  • Expressed in Escherichia coli for high yield production.
  • Lyophilized formulation for extended stability and storage.
  • Purity greater than 95% as assessed by reducing SDS-PAGE.
  • Approximate molecular weight of 26 kDa.
  • Suitable for biochemical assays and antibody validation.