Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GSTM2 protein, human (N-His) | ≥98.0% | 50 UG
SDP

Catalog No. 5000293607
Encompass_Preferred

Recombinant full-length human Glutathione S-transferase Mu 2 (GSTM2) with an N-terminal 6xHis tag, expressed in E. coli and supplied lyophilized for research applications. Characterized for purity, activity, and low endotoxin to support biochemical and immunological assays.

  • Recombinant full-length human protein with N-terminal 6xHis tag.
  • Expressed in E. coli expression system.
  • Lyophilized formulation with trehalose and Tween-80 for stability.
  • Purity ≥98.0% by reducing SDS-PAGE.
  • Bioactivity 10-15 U/mg (CDNB substrate assay).
  • Endotoxin level <1 EU/μg.
  • Not recommended to reconstitute below 100 μg/mL.

Catalog No. 50-002-93607 Supplier Medchemexpress LLC Supplier No. HYP75152A50UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant full-length human Glutathione S-transferase Mu 2 (GSTM2) with an N-terminal 6xHis tag, expressed in E. coli and supplied lyophilized for research applications. Characterized for purity, activity, and low endotoxin to support biochemical and immunological assays.

  • Recombinant full-length human protein with N-terminal 6xHis tag.
  • Expressed in E. coli expression system.
  • Lyophilized formulation with trehalose and Tween-80 for stability.
  • Purity ≥98.0% by reducing SDS-PAGE.
  • Bioactivity 10-15 U/mg (CDNB substrate assay).
  • Endotoxin level <1 EU/μg.
  • Not recommended to reconstitute below 100 μg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.