Learn More
ABclonal Technology GSTM5 Rabbit pAb
SDP
Catalog No. 5000108729
Encompass_Preferred
This gene encodes a glutathione S-transferase belonging to the mu class, part of a family of enzymes involved in detoxifying electrophilic compounds, including carcinogens, drugs, toxins, and oxidative stress products. These enzymes function by conjugating harmful substances with glutathione. The mu class genes are clustered on chromosome 1p13.3 and are highly polymorphic, which can affect an individuals susceptibility to toxins and influence the toxicity and efficacy of certain drugs. Genetic variations in the substrate-binding domains and tissue expression patterns have allowed for the diversification of these genes to handle an increasing number of foreign compounds.
Catalog No. 50-001-08729 Supplier ABclonal Technology Supplier No. A15677500UL
This gene encodes a glutathione S-transferase belonging to the mu class, part of a family of enzymes involved in detoxifying electrophilic compounds, including carcinogens, drugs, toxins, and oxidative stress products. These enzymes function by conjugating harmful substances with glutathione. The mu class genes are clustered on chromosome 1p13.3 and are highly polymorphic, which can affect an individuals susceptibility to toxins and influence the toxicity and efficacy of certain drugs. Genetic variations in the substrate-binding domains and tissue expression patterns have allowed for the diversification of these genes to handle an increasing number of foreign compounds.