Learn More
Medchemexpress LLC HY-112534 5mg Medchemexpress, GSTO-IN-2 CAS:1202710-57-3 Purity:>98%
SDP
Catalog No. 501969100
Encompass_Preferred
Medchemexpress, HY-112534 5mg GSTO-IN-2 CAS:1202710-57-3 GSTO-IN-2 is a glutathione S-transferase inhibitor with IC50s of 3.6, 16.3, and 1.4 μM for GSTA2, GSTM1, and GSTP1-1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9100 Supplier Medchemexpress LLC Supplier No. HY1125345MG
Medchemexpress, HY-112534 5mg GSTO-IN-2 CAS:1202710-57-3 GSTO-IN-2 is a glutathione S-transferase inhibitor with IC50s of 3.6, 16.3, and 1.4 μM for GSTA2, GSTM1, and GSTP1-1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.