Learn More
Medchemexpress LLC GSTO1-IN-1 | 568544-03-6 | 99.1% | 311.18 | 1 MG
SDP
Catalog No. 501968902
Encompass_Preferred
GSTO1-IN-1 is a potent inhibitor of glutathione S-transferase omega 1 (GSTO1) with an IC50 of 31 nM. It is intended for research use only. In cellular studies, it inhibits GSTO1 enzyme activity, decreases HCT116 cell viability dose-dependently, and inhibits clonogenic survival at sub-micromolar concentrations. In in vivo studies, it significantly inhibited tumor growth in a human colon cancer cell line xenograft model in nude mice without overt signs of toxicity or significant variations in average body weights over a 5-week treatment period.
  • Potent GSTO1 inhibitor with an IC50 of 31 nM.
  • Inhibits GSTO1 enzyme activity.
  • Decreases HCT116 cell viability.
  • Inhibits clonogenic survival.
  • Significantly inhibits tumor growth in xenograft models.
  • Well tolerated in in vivo studies.
Catalog No. 50-196-8902 Supplier Medchemexpress LLC Supplier No. HY11153010MG
GSTO1-IN-1 is a potent inhibitor of glutathione S-transferase omega 1 (GSTO1) with an IC50 of 31 nM. It is intended for research use only. In cellular studies, it inhibits GSTO1 enzyme activity, decreases HCT116 cell viability dose-dependently, and inhibits clonogenic survival at sub-micromolar concentrations. In in vivo studies, it significantly inhibited tumor growth in a human colon cancer cell line xenograft model in nude mice without overt signs of toxicity or significant variations in average body weights over a 5-week treatment period.
  • Potent GSTO1 inhibitor with an IC50 of 31 nM.
  • Inhibits GSTO1 enzyme activity.
  • Decreases HCT116 cell viability.
  • Inhibits clonogenic survival.
  • Significantly inhibits tumor growth in xenograft models.
  • Well tolerated in in vivo studies.