Learn More
ABclonal Technology GSTO1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A15354500UL
Encompass_Preferred
The protein encoded by this gene is an omega class glutathione S-transferase (GST) with glutathione-dependent thiol transferase and dehydroascorbate reductase activities. GSTs are involved in the metabolism of xenobiotics and carcinogens. The encoded protein acts as a homodimer and is found in the cytoplasm. Three transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-05136