Learn More
Aviva Systems Biology Corp GSTO2 Antibody - middle region (ARP49169_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49169_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GSTO2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle of human GSTO2. Alternate names: bA127L20.1, GSTO 2-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-60034