Learn More
Aviva Systems Biology Corp GSTP1 Antibody - N-terminal region (ARP48487_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48487_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GSTP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human GSTP1. Alternate names: DFN7, FAEES3, GST3, GSTP, PIApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53931