Learn More
Sigma Aldrich Fine Chemicals Biosciences GSTrapTM fast flow Cytiva
Catalog No. 502904696
Encompass
GSTrap(TM) ff columns are prepacked Glutathione Sepharose(R) fast fLow columns for fast convenient one-step purification of glutathione S-transferase (GST) tagged proteins.
Catalog No. 50-290-4696 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17513101
GSTrap(TM) ff columns are prepacked Glutathione Sepharose(R) fast fLow columns for fast convenient one-step purification of glutathione S-transferase (GST) tagged proteins.