Learn More
Sigma Aldrich Fine Chemicals Biosciences GSTrapTM High Performance
Catalog No. 502904454
Encompass
GSTrap(TM) HP columns are prepacked with Glutathione Sepharose(R) High Performance medium and provide high-reSOLUTIONon one-step purifications of glutathione S-Transferase (GST) tagged proteins.
Catalog No. 50-290-4454 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17528201
Add to Cart
Add to Cart
GSTrap(TM) HP columns are prepacked with Glutathione Sepharose(R) High Performance medium and provide high-reSOLUTIONon one-step purifications of glutathione S-Transferase (GST) tagged proteins.