Learn More
ABclonal Technology GSTT2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A15678500UL
Encompass_Preferred
The protein encoded by this gene, glutathione S-transferase (GST) theta 2 (GSTT2), is a member of a superfamily of proteins that catalyze the conjugation of reduced glutathione to a variety of electrophilic and hydrophobic compounds. Human GSTs can be divided into five main classes alpha, mu, pi, theta, and zeta. The theta class includes GSTT1, GSTT2, and GSTT2B. GSTT2 and GSTT2B are nearly identical to each other, and share 55% amino acid identity with GSTT1. All three genes may play a role in human carcinogenesis. The GSTT2 gene is a pseudogene in some populations.
Catalog No. 50-001-08116