Learn More
Medchemexpress LLC Gt-653 | 3057627-63-8 | 99.8% | 653.60 | C32H27N7O9 | 10 MG
SDP
Catalog No. 5000288318
Encompass_Preferred
GT-653 is a PROTAC degrader that targets lysine-specific demethylase 5B (KDM5B), promoting ubiquitin-proteasome-dependent degradation and modulating H3K4 methylation in cellular models. It is supplied as a white to off-white solid for research use in biochemical and cellular studies of epigenetic regulation.
  • PROTAC mechanism targeting KDM5B, a histone demethylase.
  • Demonstrated 68.35% degradation of KDM5B at 10 μM in cell assays.
  • Upregulates H3K4me3 levels and activates type-I interferon signaling in prostate cancer cells.
  • Available as a white to off-white powder suitable for in vitro studies.
  • Molecular formula C32H27N7O9; molecular weight 653.60.
Catalog No. 50-002-88318 Supplier Medchemexpress LLC Supplier No. HY15843210MG
GT-653 is a PROTAC degrader that targets lysine-specific demethylase 5B (KDM5B), promoting ubiquitin-proteasome-dependent degradation and modulating H3K4 methylation in cellular models. It is supplied as a white to off-white solid for research use in biochemical and cellular studies of epigenetic regulation.
  • PROTAC mechanism targeting KDM5B, a histone demethylase.
  • Demonstrated 68.35% degradation of KDM5B at 10 μM in cell assays.
  • Upregulates H3K4me3 levels and activates type-I interferon signaling in prostate cancer cells.
  • Available as a white to off-white powder suitable for in vitro studies.
  • Molecular formula C32H27N7O9; molecular weight 653.60.