Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GT 949 100mg | 460330-27-2 | 100 MG
SDP

Catalog No. 5000229042
Encompass_Preferred

GT 949 is a selective excitatory amino acid transporter-2 (EAAT2) positive allosteric modulator that enhances glutamate uptake noncompetitively. It is highly potent and selective for EAAT2, supplied as a high-purity solid for research use.

  • Positive allosteric modulator of EAAT2.
  • Potent activity with EC50 of 0.26 nM.
  • Selective for EAAT2 versus EAAT1 and EAAT3.
  • High purity (99.55%) solid, off-white to light yellow.
  • Molecular formula C30H37N7O2; molecular weight 527.66 g·mol⁻1.
  • Recommended storage: powder -20°C for long term, 4°C for short term.

Catalog No. 50-002-29042 Supplier Medchemexpress LLC Supplier No. HY114381100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GT 949 is a selective excitatory amino acid transporter-2 (EAAT2) positive allosteric modulator that enhances glutamate uptake noncompetitively. It is highly potent and selective for EAAT2, supplied as a high-purity solid for research use.

  • Positive allosteric modulator of EAAT2.
  • Potent activity with EC50 of 0.26 nM.
  • Selective for EAAT2 versus EAAT1 and EAAT3.
  • High purity (99.55%) solid, off-white to light yellow.
  • Molecular formula C30H37N7O2; molecular weight 527.66 g·mol⁻1.
  • Recommended storage: powder -20°C for long term, 4°C for short term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.