Learn More
Medchemexpress LLC GT 949 25mg | 460330-27-2 | 25 MG
SDP
Catalog No. 5000229038
Encompass_Preferred
GT 949 is a selective positive allosteric modulator of the excitatory amino acid transporter 2 (EAAT2) that enhances glutamate transport with high potency (EC50 = 0.26 nM). It is supplied as a research reagent and is stable under recommended storage conditions.
  • Selective EAAT2 positive allosteric modulator with EC50 ≈ 0.26 nM.
  • Enhances glutamate transport without affecting EAAT1 or EAAT3.
  • High purity suitable for research applications.
  • Molecular formula C30H37N7O2; molecular weight 527.66 g·mol⁻1.
  • Store powder at -20°C; store in solution at -80°C for long-term storage.
Catalog No. 50-002-29038 Supplier Medchemexpress LLC Supplier No. HY11438125MG
GT 949 is a selective positive allosteric modulator of the excitatory amino acid transporter 2 (EAAT2) that enhances glutamate transport with high potency (EC50 = 0.26 nM). It is supplied as a research reagent and is stable under recommended storage conditions.
  • Selective EAAT2 positive allosteric modulator with EC50 ≈ 0.26 nM.
  • Enhances glutamate transport without affecting EAAT1 or EAAT3.
  • High purity suitable for research applications.
  • Molecular formula C30H37N7O2; molecular weight 527.66 g·mol⁻1.
  • Store powder at -20°C; store in solution at -80°C for long-term storage.