Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc Gt aGuinea Pig IgG Anti

Catalog No. 5000087223
Encompass_Preferred

Product Name: HRP Goat Anti-Guinea Pig IgG (H&L)
Product Class: Antibody
Type: Secondary
Product Name: HRP Goat Anti-Guinea Pig IgG (H&L)
Cytek Catalog Number: 72-8098-M001
Marker: IgG (H&L)
Target Species: Guinea Pig
Reported Cross Reactivity: Guinea Pig
Clonality: Polyclonal
Host Species: Goat
Conjugation: HRP
Size: 1 mg

Catalog No. 50-000-87223 Supplier Cytek Biosciences Inc Supplier No. 728098M001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: HRP Goat Anti-Guinea Pig IgG (H&L)
Product Class: Antibody
Type: Secondary
Product Name: HRP Goat Anti-Guinea Pig IgG (H&L)
Cytek Catalog Number: 72-8098-M001
Marker: IgG (H&L)
Target Species: Guinea Pig
Reported Cross Reactivity: Guinea Pig
Clonality: Polyclonal
Host Species: Goat
Conjugation: HRP
Size: 1 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.