Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc Gt aMu IgG2a PE Anti

Catalog No. 5000086596
Encompass_Preferred

Product Name: PE Goat Anti-Mouse IgG2a
Product Class: Antibody
Type: Secondary
Product Name: PE Goat Anti-Mouse IgG2a
Cytek Catalog Number: 50-8116-U100
Marker: IgG2a
Target Species: Mouse
Reported Cross Reactivity: Mouse
Clonality: Polyclonal
Host Species: Goat
Conjugation: PE
Size: 100 μg

Catalog No. 50-000-86596 Supplier Cytek Biosciences Inc Supplier No. 508116U100
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: PE Goat Anti-Mouse IgG2a
Product Class: Antibody
Type: Secondary
Product Name: PE Goat Anti-Mouse IgG2a
Cytek Catalog Number: 50-8116-U100
Marker: IgG2a
Target Species: Mouse
Reported Cross Reactivity: Mouse
Clonality: Polyclonal
Host Species: Goat
Conjugation: PE
Size: 100 μg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.