Learn More
Immunology Consultants anti-Rat Alpha 2-Macroglobulin Antibody (Goat) - Affinity Purified
SDP
Catalog No. 502248782
Encompass_Preferred
This affinity‑purified goat antibody provides sensitive, specific detection of rat Alpha 2‑Macroglobulin, a major plasma protease inhibitor involved in inflammation, coagulation, and acute‑phase responses. Its strong affinity and low nonspecific binding support reliable performance in ELISA, Western blotting, and immunoassay development. The antibody is well suited for studies of liver function, systemic inflammation, and biomarker profiling in rat models. The 0.1 mg format is ideal for pilot assays, limited‑volume workflows, and method optimization requiring consistent, high‑quality detection of Alpha 2‑Macroglobulin.
This item is not returnable. View return policy
Catalog No. 50-224-8782 Supplier Immunology Consultants Supplier No. G2M25AZ
This affinity‑purified goat antibody provides sensitive, specific detection of rat Alpha 2‑Macroglobulin, a major plasma protease inhibitor involved in inflammation, coagulation, and acute‑phase responses. Its strong affinity and low nonspecific binding support reliable performance in ELISA, Western blotting, and immunoassay development. The antibody is well suited for studies of liver function, systemic inflammation, and biomarker profiling in rat models. The 0.1 mg format is ideal for pilot assays, limited‑volume workflows, and method optimization requiring consistent, high‑quality detection of Alpha 2‑Macroglobulin.