Learn More
Biocare Medical, LLC GT-ON-RODENT AP-POLYMER 25ML
SDP
Catalog No. 5082861
Encompass_Preferred
Goat-on-Rodent AP-Polymer is for goat primary antibodies on mouse, rat or human tissues and labeled with alkaline phosphatase. Goat antibodies are advantageous as the detection hass minimum cross reactivity to mouse, rat or human IgG.
Catalog No. 50-828-61 Supplier Biocare Medical, LLC Supplier No. GAP514H
Goat-on-Rodent AP-Polymer is for goat primary antibodies on mouse, rat or human tissues and labeled with alkaline phosphatase. Goat antibodies are advantageous as the detection hass minimum cross reactivity to mouse, rat or human IgG.