Learn More
Biocare Medical, LLC Goat-on-Rodent HRP-Polymer, 100 ml
SDP
Catalog No. 5082864
Encompass_Preferred
Goat-on-Rodent HRP-Polymer is for goat primary antibodies on mouse, rat or human tissues and labeled with horseradish peroxidase. Goat antibodies are advantageous as the detection has minimum cross reactivity to mouse, rat or human IgG.
Catalog No. 50-828-64 Supplier Biocare Medical, LLC Supplier No. GHP516L
Goat-on-Rodent HRP-Polymer is for goat primary antibodies on mouse, rat or human tissues and labeled with horseradish peroxidase. Goat antibodies are advantageous as the detection has minimum cross reactivity to mouse, rat or human IgG.