Learn More
Enzo Life Sciences Ganglioside GT1b. trisodium salt (bovine brain) (1mg). CAS: 59247-13-1
SDP
Supplier:  Enzo Life Sciences ALX302011M001
Encompass_Preferred
Prevents neurotoxicity of glutamate on neuronal cells. Alternative name: GT1b . 3Na (bovine brain), Trisialoganglioside GT1b . 3Na (bovine brain). Purity: ≥98% (TLC). Solubility: Very soluble in water, biological buffers, 100% ethanol or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8651