Learn More
StatLab™ GTF™ Formalin Substitute
SDP
Catalog No. STLFS1
Change view
Click to view available options
Quantity:
4 x 1 gal.
Catalog No. Quantity
STLFS1 4 x 1 gal.
1 options
This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.
Catalog No. STLFS1 Supplier StatLab™ Supplier No. FS1
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.
A glyoxal based, formalin-free fixative ideal for conventional histology, special stains and immunohistochemistry.
  • Formaldehyde-free solution provides morphological detail of standard aldehyde fixative while preserving high levels of immunoreactivity
Specifications
Packaging Type Bottle
Quantity 4 x 1 gal.
For Use With (Application) Tissue Fixation