Learn More
Aviva Systems Biology Corp GTF2A1 Antibody - middle region (ARP58156_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58156_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GTF2A1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human GTF2A1. Alternate names: MGC129969, MGC129970, TF2A1, TFIIAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Yeast
Catalog No. 50-286-445