Learn More
Aviva Systems Biology Corp GTF2A1L Antibody - C-terminal region (ARP53837_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53837_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GTF2A1L. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human GTF2A1L. Alternate names: ALF, MGC26254ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rat
Catalog No. 50-285-895