Learn More
Aviva Systems Biology Corp GTF2E2 Antibody - N-terminal region (ARP31434_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31434_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GTF2E2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GTF2E2. Alternate names: FE, TF2E2, TFIIE-BApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54219