Learn More
Aviva Systems Biology Corp GTF2F2 Antibody - C-terminal region (ARP31436_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31436_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GTF2F2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human GTF2F2. Alternate names: BTF4, RAP30, TF2F2, TFIIFApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54220