Learn More
Aviva Systems Biology Corp GTF2H1 Antibody - middle region (P100929_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100929_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GTF2H1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GTF2H1. Alternate names: BTF2, TFIIH, TFB1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-62788