Learn More
Aviva Systems Biology Corp GTF2H2 Antibody - C-terminal region (P100894_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100894_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GTF2H2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human GTF2H2. Alternate names: p44, BTF2, TFIIH, BTF2P44, T-BTF2P44ApplicationIHC, WB. Species reactivity: Dog, Horse, Human, Mouse, Zebrafish
Catalog No. 50-014-62771