Learn More
Aviva Systems Biology Corp GTF2H2 Antibody - middle region (P100894_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100894_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GTF2H2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GTF2H2. Alternate names: BTF2, BTF2P44, MGC102806, T-BTF2P44, TFIIH, p44ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Zebrafish
Catalog No. 50-014-62770