Learn More
Aviva Systems Biology Corp Gtf2h3 Antibody - N-terminal region (ARP37596_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37596_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Gtf2h3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 34kDa, 5033417D07Rik, BTF2, C730029A10, D5Ertd679e, TFIIH, BTF2 p34ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-665