Learn More
Aviva Systems Biology Corp GTF2H3 Antibody - N-terminal region (ARP31438_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31438_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GTranscription Factor Antibodies2H3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human GTF2H3. Alternate names: BTF2, TFB4, TFIIHApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54223