Learn More
Aviva Systems Biology Corp GTF2H4 Antibody - N-terminal region (ARP32598_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32598_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GTF2H4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GTF2H4. Alternate names: TFIIH, TFB2ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-54670