Learn More
Aviva Systems Biology Corp GTF2I Antibody - N-terminal region (ARP31910_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31910_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GTF2I. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GTF2I. Alternate names: WBS, DIWS, SPIN, IB291, BAP135, BTKAP1, TFII-I, WBSCR6, GTFII-IApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54367