Learn More
Aviva Systems Biology Corp GTF2I Antibody - middle region (ARP58007_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58007_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GTF2I. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GTF2I. Alternate names: BAP-135, BAP135, BTKAP1, DIWS, IB291, SPIN, TFII-I, WBS, WBSCR6, GTFII-IApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62211