Learn More
Aviva Systems Biology Corp GTF2IRD1 Antibody - C-terminal region (ARP39254_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39254_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GTF2IRD1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human GTF2IRD1. Alternate names: CREAM1, GTF3, MUSTRD1, RBAP2, WBSCR11, WBSCR12, hMusTRD1alpha1, BEN, WBSApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-56960