Learn More
Aviva Systems Biology Corp GTF3C3 Antibody - N-terminal region (ARP35841_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35841_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GTF3C3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GTF3C3. Alternate names: TFIIIC102, TFIIICgamma, TFiiiC2-102ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-55887