Learn More
Aviva Systems Biology Corp CFAP20 Antibody - C-terminal region (ARP34301_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34301_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GTL3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GTL3. Alternate names: GTL3, EVORF, fSAP23ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-52949