Learn More
Cytoskeleton, Inc. GTP 10X 100UL
Catalog No. NC9896726
Encompass
GTP,Buffers used for Tubulin Research,100mM solution of GTP,Required for tubulin stability and microtubule assembly and dynamics,Amount:10x100ul
Catalog No. NC9896726 Supplier Cytoskeleton, Inc. Supplier No. BST06010
GTP,Buffers used for Tubulin Research,100mM solution of GTP,Required for tubulin stability and microtubule assembly and dynamics,Amount:10x100ul