Learn More
Medchemexpress LLC GTP-14564 | 34823-86-4 | 99.9% | 234.25 | 100 MG
SDP
Catalog No. 5000344077
Encompass_Preferred
GTP-14564 is a tyrosine kinase inhibitor that targets internal tandem duplication (ITD) and FLT3. It inhibits FLT3 ligand-dependent growth in Ba/F3 leukemia cells.
  • Tyrosine kinase inhibitor
  • Targets internal tandem duplication (ITD) and FLT3
  • Inhibits FLT3 ligand-dependent growth in Ba/F3 leukemia cells
  • For research use only
Catalog No. 50-003-44077 Supplier Medchemexpress LLC Supplier No. HY11002610MM/1ML
GTP-14564 is a tyrosine kinase inhibitor that targets internal tandem duplication (ITD) and FLT3. It inhibits FLT3 ligand-dependent growth in Ba/F3 leukemia cells.
  • Tyrosine kinase inhibitor
  • Targets internal tandem duplication (ITD) and FLT3
  • Inhibits FLT3 ligand-dependent growth in Ba/F3 leukemia cells
  • For research use only