Learn More
Medchemexpress LLC GTP-14564 | 34823-86-4 | 99.9% | 234.25 | 50 MG
SDP
Catalog No. 5000344818
Encompass_Preferred
GTP-14564 is a tyrosine kinase inhibitor that targets internal tandem duplication (ITD) and FLT3. It inhibits FLT3 ligand-dependent growth in Ba/F3 leukemia cells. It is a potential research tool for studying FLT3-related leukemia.
  • Tyrosine kinase inhibitor
  • Targets internal tandem duplication (ITD)
  • Targets FLT3
  • Inhibits FLT3 ligand-dependent growth in Ba/F3 leukemia cells
  • Appearance: solid
  • Color: off-white to yellow
Catalog No. 50-003-44818 Supplier Medchemexpress LLC Supplier No. HY11002610MG
GTP-14564 is a tyrosine kinase inhibitor that targets internal tandem duplication (ITD) and FLT3. It inhibits FLT3 ligand-dependent growth in Ba/F3 leukemia cells. It is a potential research tool for studying FLT3-related leukemia.
  • Tyrosine kinase inhibitor
  • Targets internal tandem duplication (ITD)
  • Targets FLT3
  • Inhibits FLT3 ligand-dependent growth in Ba/F3 leukemia cells
  • Appearance: solid
  • Color: off-white to yellow